Assay Details
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | ACUGCUGAGCUAGCACUUCCCG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-93-3p | MIMAT0004509 |
| Mouse | mmu-miR-93-3p | MIMAT0004636 |
| Pteropus alecto | pal-miR-93-3p | MIMAT0039938 |
| Rhesus monkey | mml-miR-93-3p | MIMAT0026602 |
Stem-loop Details
Human
Mouse
Pteropus alecto
Rhesus monkey
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Human Pool B v3.0Megaplex™ RT Primers, Rodent Pool B v3.0
Associated TaqMan® Array MicroRNA Cards:
TaqMan® Human MicroRNA B Cards v3.0TaqMan® Rodent MicroRNA B Cards v3.0
TaqMan® OpenArray® MicroRNA Panels:
TaqMan® OpenArray® Human MicroRNA PanelTaqMan® OpenArray® Rodent MicroRNA Panel
Related Products
miRBase Alias:
Human: hsa-miR-93* (v17)
Mouse: mmu-miR-93* (v17)
Mouse: mmu-miR-93* (v17)

Genomic Map