Assay Details
Amplicon Length: | 56 |
Chromosome Location: | Chr. 7 - 100093414 - 100093414 [-] on 100093414 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-25-5p | MIMAT0004498 |
| Human | hsa-miR-25-3p | MIMAT0000081 |
| Human | hsa-miR-93-5p | MIMAT0000093 |
| Human | hsa-miR-93-3p | MIMAT0004509 |
Stem-loop Details
Human

Genomic Map