Assay Details
Gene Family ID: | MIPF0000513, mir-924 |
Mature miRNA Sequence: | AGAGUCUUGUGAUGUCUUGC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-924 | MIMAT0004974 |
| Pan troglodytes | ptr-miR-924 | MIMAT0008339 |
| Pongo pygmaeus | ppy-miR-924 | MIMAT0016133 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus

Genomic Map