Assay Details
Amplicon Length: | 77 |
Chromosome Location: | Chr. 18 - 39621928 - 39621928 [-] on 39621928 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-924 | MIMAT0004974 |
Stem-loop Details
Human
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amplicon Length: | 77 |
Chromosome Location: | Chr. 18 - 39621928 - 39621928 [-] on 39621928 Build GRCh38 |
Species: | Human |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-924 | MIMAT0004974 |