Assay Details
Gene Family ID: | MIPF0000386, mir-743 |
Mature miRNA Sequence: | CACUGUGUCCUUUCUGCGUAG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-892a | MIMAT0004907 |
| Pan troglodytes | ptr-miR-892a | MIMAT0008335 |
| Rhesus monkey | mml-miR-892a | MIMAT0006531 |
Stem-loop Details
Human
Pan troglodytes
Rhesus monkey

Genomic Map