Assay Details
Amplicon Length: | 101 |
Chromosome Location: | Chr. X - 145997002 - 145997002 [+] on 145997002 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-892a | MIMAT0004907 |
| Human | hsa-miR-892b | MIMAT0004918 |
Stem-loop Details
Human

Genomic Map