Assay Details
Gene Family ID: | MIPF0000386, mir-743 |
Mature miRNA Sequence: | CACUGGCUCCUUUCUGGGUAGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-892b | MIMAT0004918 |
| Pongo pygmaeus | ppy-miR-892b | MIMAT0016129 |
| Rhesus monkey | mml-miR-892b | MIMAT0012783 |
Stem-loop Details
Human
Pongo pygmaeus
Rhesus monkey

Genomic Map