Assay Details
Gene Family ID: | MIPF0000190, mir-342 |
Mature miRNA Sequence: | UCUCACACAGAAAUCGCACCCGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-342-3p | MIMAT0000753 | |
| Mouse | mmu-miR-342-3p | MIMAT0000590 | |
| Rat | rno-miR-342-3p | MIMAT0000589 | |
| Dog | cfa-miR-342 | MIMAT0006709 | |
| Horse | eca-miR-342-3p | MIMAT0013137 | |
| Pan troglodytes | ptr-miR-342 | MIMAT0008110 | |
| View More Rows | |||
| Pongo pygmaeus | ppy-miR-342-3p | MIMAT0015845 | |
| Pteropus alecto | pal-miR-342-3p | MIMAT0040086 | |
| Rhesus monkey | mml-miR-342-3p | MIMAT0006283 | |
| Tupaia chinensis | tch-miR-342-3p | MIMAT0036487 | |
Stem-loop Details
Human
Mouse
Rat
Dog
Horse
Pan troglodytes
Pongo pygmaeus
Pteropus alecto
Rhesus monkey
Tupaia chinensis
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Human Pool AMegaplex™ RT Primers, Rodent Pool A
Related Products

Genomic Map