Assay Details
Gene Family ID: | MIPF0000190, mir-342 |
Mature miRNA Sequence: | UCUCACACAGAAAUCGCACCCGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-342-3p | MIMAT0000753 |
| Mouse | mmu-miR-342-3p | MIMAT0000590 |
| Rat | rno-miR-342-3p | MIMAT0000589 |
| Dog | cfa-miR-342 | MIMAT0006709 |
| Horse | eca-miR-342-3p | MIMAT0013137 |
| Pan troglodytes | ptr-miR-342 | MIMAT0008110 |
| View More Rows | ||
| Pongo pygmaeus | ppy-miR-342-3p | MIMAT0015845 |
| Pteropus alecto | pal-miR-342-3p | MIMAT0040086 |
| Rhesus monkey | mml-miR-342-3p | MIMAT0006283 |
| Tupaia chinensis | tch-miR-342-3p | MIMAT0036487 |
Stem-loop Details
Human
Mouse
Rat
Dog
Horse
Pan troglodytes
Pongo pygmaeus
Pteropus alecto
Rhesus monkey
Tupaia chinensis

Genomic Map