Assay Details
Gene Family ID: | MIPF0000395, mir-760 |
Mature miRNA Sequence: | CGGCUCUGGGUCUGUGGGGA |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Human | hsa-miR-760 | MIMAT0004957 | |
Mouse | mmu-miR-760-3p | MIMAT0003898 | |
Rat | rno-miR-760-3p | MIMAT0005337 | |
Bovine | bta-miR-760-3p | MIMAT0022951 | |
Cricetulus griseus | cgr-miR-760 | MIMAT0041166 | |
Pan troglodytes | ptr-miR-760 | MIMAT0008319 | |
View More Rows | |||
Pongo pygmaeus | ppy-miR-760 | MIMAT0016104 | |
Rhesus monkey | mml-miR-760 | MIMAT0012788 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Cricetulus griseus
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
