Assay Details
Gene Family ID: | MIPF0000395, mir-760 |
Mature miRNA Sequence: | CGGCUCUGGGUCUGUGGGGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-760 | MIMAT0004957 |
| Mouse | mmu-miR-760-3p | MIMAT0003898 |
| Rat | rno-miR-760-3p | MIMAT0005337 |
| Bovine | bta-miR-760-3p | MIMAT0022951 |
| Cricetulus griseus | cgr-miR-760 | MIMAT0041166 |
| Pan troglodytes | ptr-miR-760 | MIMAT0008319 |
| View More Rows | ||
| Pongo pygmaeus | ppy-miR-760 | MIMAT0016104 |
| Rhesus monkey | mml-miR-760 | MIMAT0012788 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Cricetulus griseus
Pan troglodytes
Pongo pygmaeus
Rhesus monkey

Genomic Map