Assay Details
Gene Family ID: | MIPF0000002, let-7 |
Mature miRNA Sequence: | CUAUACAGUCUACUGUCUUUCC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-let-7f-2-3p | MIMAT0004487 |
| Dasypus novemcinctus | dno-let-7f-2-3p | MIMAT0047484 |
| Guinea pig | cpo-let-7f-2-3p | MIMAT0046830 |
| Monodelphis domestica | mdo-let-7f-2-3p | MIMAT0031040 |
| Rabbit | ocu-let-7f-2-3p | MIMAT0048086 |
| Salmo salar | ssa-let-7g-3p | MIMAT0032271 |
Stem-loop Details
Human
Dasypus novemcinctus
Guinea pig
Monodelphis domestica
Rabbit
Salmo salar

Genomic Map