Assay Details
Amplicon Length: | 62 |
Chromosome Location: | Chr. X - 53556986 - 53556986 [+] on 53556986 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-let-7f-2-3p | MIMAT0004487 |
| Human | hsa-let-7f-5p | MIMAT0000067 |
Stem-loop Details
Human

Genomic Map