Assay Details
Gene Family ID: | MIPF0000173, mir-499 |
Mature miRNA Sequence: | AACAUCACAGCAAGUCUGUGCU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-499a-3p | MIMAT0004772 | |
| Rat | rno-miR-499-3p | MIMAT0017227 | |
| Dasypus novemcinctus | dno-miR-499-3p | MIMAT0047937 | |
| Guinea pig | cpo-miR-499-3p | MIMAT0047298 | |
| Horse | eca-miR-499-3p | MIMAT0013109 | |
| Pan troglodytes | ptr-miR-499 | MIMAT0008172 | |
| View More Rows | |||
| Pig | ssc-miR-499-3p | MIMAT0017373 | |
| Pongo pygmaeus | ppy-miR-499-3p | MIMAT0015921 | |
| Pteropus alecto | pal-miR-499a-3p | MIMAT0039821 | |
| Rabbit | ocu-miR-499-3p | MIMAT0048490 | |
| Rhesus monkey | mml-miR-499-3p | MIMAT0006362 | |
Stem-loop Details
Human
Rat
Dasypus novemcinctus
Guinea pig
Horse
Pan troglodytes
Pig
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-499-3p (v17)
Rat: rno-miR-499* (v18)
Rat: rno-miR-499* (v18)

Genomic Map