Assay Details
Amplicon Length: | 67 |
Chromosome Location: | Chr. 20 - 34990680 - 34990680 [+] on 34990680 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-499a-5p | MIMAT0002870 |
| Human | hsa-miR-499a-3p | MIMAT0004772 |
| Human | hsa-miR-499b-3p | MIMAT0019898 |
| Human | hsa-miR-499b-5p | MIMAT0019897 |
Stem-loop Details
Human

Genomic Map