Assay Details
Gene Family ID: | MIPF0000068, mir-290 |
Mature miRNA Sequence: | ACUCAAACUGUGUGACAUUUUG |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-293-5p | MIMAT0004573 |
Stem-loop Details
Mouse

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | mmu-miR-293* |
---|---|
miRBase Accession Number: | MI0000391 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | ACUCAAACUGUGUGACAUUUUG |
Species: | Mouse |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000068, mir-290 |
Mature miRNA Sequence: | ACUCAAACUGUGUGACAUUUUG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-293-5p | MIMAT0004573 |