Assay Details
Amplicon Length: | 62 |
Chromosome Location: | Chr. 7 - 3220144 - 3220144 [-] on 3220144 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-293-3p | MIMAT0000371 |
Mouse | mmu-miR-293-5p | MIMAT0004573 |
Stem-loop Details
Mouse
