Assay Details
Gene Family ID: | MIPF0000128, mir-450 |
Mature miRNA Sequence: | AUUGGGAACAUUUUGCAUGCAU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-450b-3p | MIMAT0003512 |
| Rabbit | ocu-miR-450a-3p | MIMAT0048458 |
Stem-loop Details
Mouse
Rabbit

Genomic Map