Assay Details
Amplicon Length: | 96 |
Chromosome Location: | Chr. X - 53047749 - 53047749 [+] on 53047749 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-450b-5p | MIMAT0003511 |
| Mouse | mmu-miR-450b-3p | MIMAT0003512 |
Stem-loop Details
Mouse

Genomic Map