Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | AAAGUGCUUCUCUUUGGUGGGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-520d-3p | MIMAT0002856 |
| Pan troglodytes | ptr-miR-520d | MIMAT0008200 |
| Pongo pygmaeus | ppy-miR-520d-3p | MIMAT0015972 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus

Genomic Map