Assay Details
Amplicon Length: | 69 |
Chromosome Location: | Chr. 19 - 53719895 - 53719895 [-] on 53719895 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-520d-5p | MIMAT0002855 |
| Human | hsa-miR-520d-3p | MIMAT0002856 |
Stem-loop Details
Human

Genomic Map