Assay Details
Gene Family ID: | MIPF0000445, mir-1225 |
Mature miRNA Sequence: | GUGGGUACGGCCCAGUGGGGGG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1225-5p | MIMAT0005572 |
| Pongo pygmaeus | ppy-miR-1225-5p | MIMAT0016160 |
Stem-loop Details
Human
Pongo pygmaeus

Genomic Map