Assay Details
Amplicon Length: | 60 |
Chromosome Location: | Chr. 16 - 2090597 - 2090597 [-] on 2090597 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-1225-3p | MIMAT0005573 |
Human | hsa-miR-1225-5p | MIMAT0005572 |
Stem-loop Details
Human
