Assay Details
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | AAAAGUACUUGCGGAUUUUGCU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-548k | MIMAT0005882 |
Pan troglodytes | ptr-miR-548k | MIMAT0008225 |
Stem-loop Details
Human
Pan troglodytes
