Assay Details
Amplicon Length: | 88 |
Chromosome Location: | Chr. 11 - 70284261 - 70284261 [+] on 70284261 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-548k | MIMAT0005882 |
Stem-loop Details
Human
