Assay Details
Gene Family ID: | MIPF0000320, mir-428 |
Mature miRNA Sequence: | UAAGUGCUCUCUAGUUCGGUUG |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Xenopus laevis | xla-miR-428 | MIMAT0001345 |
Xenopus tropicalis | xtr-miR-428a | MIMAT0003709 |
Stem-loop Details
Xenopus laevis
Xenopus tropicalis
