Assay Details
Gene Family ID: | MIPF0000320, mir-428 |
Mature miRNA Sequence: | UAAGUGCUCUCUAGUUCGGUUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Xenopus laevis | xla-miR-428 | MIMAT0001345 |
| Xenopus tropicalis | xtr-miR-428a | MIMAT0003709 |
Stem-loop Details
Xenopus laevis
Xenopus tropicalis

Genomic Map