Assay Details
Gene Family ID: | MIPF0000141, mir-315 |
Mature miRNA Sequence: | UUUUGAUUGUUGCUCAGAAAG |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Saccoglossus kowalevskii | sko-miR-315 | MIMAT0009642 |
Stem-loop Details
Saccoglossus kowalevskii
