Assay Details
Gene Family ID: | MIPF0000141, mir-315 |
Mature miRNA Sequence: | UUUUGAUUGUUGCUCAGAAAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Saccoglossus kowalevskii | sko-miR-315 | MIMAT0009642 |
Stem-loop Details
Saccoglossus kowalevskii
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | sko-miR-315 |
|---|---|
| miRBase Accession Number: | MI0010178 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUUUGAUUGUUGCUCAGAAAG |
| Species: | Saccoglossus kowalevskii |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000141, mir-315 |
Mature miRNA Sequence: | UUUUGAUUGUUGCUCAGAAAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Saccoglossus kowalevskii | sko-miR-315 | MIMAT0009642 |