Assay Details
Gene Family ID: | MIPF0000141, mir-315 |
Mature miRNA Sequence: | CUUUCGAGCAGUAAUCAAAGUC |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Aedes aegypti | aae-miR-315-3p | MIMAT0015370 |
Stem-loop Details
Aedes aegypti

More Information
Related Products
miRBase Alias:
Achromobacter aegrifaciens: aae-miR-315* (v18)
