Assay Details
Gene Family ID: | MIPF0000141, mir-315 |
Mature miRNA Sequence: | CUUUCGAGCAGUAAUCAAAGUC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Aedes aegypti | aae-miR-315-3p | MIMAT0015370 |
Stem-loop Details
Aedes aegypti
More Information
Related Products
miRBase Alias:
Achromobacter aegrifaciens: aae-miR-315* (v18)

Genomic Map