Assay Details
Mature miRNA Sequence: | UGGCACUUCAAUUGGGCUGACU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-2495-5p | MIMAT0012204 |
Stem-loop Details
Drosophila melanogaster
More Information
Related Products
miRBase Alias:
Drosophila melanogaster: dme-miR-2495 (v16)

Genomic Map