Assay Details
Mature miRNA Sequence: | UGGCACUUCAAUUGGGCUGACU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Drosophila melanogaster | dme-miR-2495-5p | MIMAT0012204 |
Stem-loop Details
Drosophila melanogaster

More Information
Related Products
miRBase Alias:
Drosophila melanogaster: dme-miR-2495 (v16)
