Assay Details
Gene Family ID: | MIPF0000088, mir-224 |
Mature miRNA Sequence: | AAAUGGUGCCCUAGUGACUACA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-224-3p | MIMAT0017062 |
| Rat | rno-miR-224-3p | MIMAT0017200 |
Stem-loop Details
Mouse
Rat
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-224* (v17)
Rat: rno-miR-224* (v18)
Rat: rno-miR-224* (v18)

Genomic Map