Assay Details
Amplicon Length: | 67 |
Chromosome Location: | Chr. X - 72260820 - 72260820 [+] on 72260820 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-224-3p | MIMAT0017062 |
| Mouse | mmu-miR-224-5p | MIMAT0000671 |
Stem-loop Details
Mouse

Genomic Map