Assay Details
Gene Family ID: | MIPF0000167, mir-370 |
Mature miRNA Sequence: | CAGGUCACGUCUCUGCAGUUAC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-370-5p | MIMAT0026483 |
| Pteropus alecto | pal-miR-370-5p | MIMAT0040053 |
| Rhesus monkey | mml-miR-370-5p | MIMAT0026857 |
| Sheep | oar-miR-370-5p | MIMAT0019243 |
Stem-loop Details
Human
Pteropus alecto
Rhesus monkey
Sheep

Genomic Map