Assay Details
Amplicon Length: | 62 |
Chromosome Location: | Chr. 14 - 100911439 - 100911439 [-] on 100911439 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-370-3p | MIMAT0000722 |
| Human | hsa-miR-370-5p | MIMAT0026483 |
Stem-loop Details
Human

Genomic Map