Assay Details
Gene Family ID: | MIPF0000186, mir-299 |
Mature miRNA Sequence: | UAUGUGGGACGGUAAACCGCU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-299a-3p | MIMAT0017167 |
| Pan paniscus | ppa-miR-299 | MIMAT0049045 |
| Papio hamadryas | pha-miR-299 | MIMAT0049578 |
Stem-loop Details
Rat
Pan paniscus
Papio hamadryas

Genomic Map