Assay Details
Amplicon Length: | 64 |
Chromosome Location: | Chr. 6 - 133859169 - 133859169 [+] on 133859169 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-299a-3p | MIMAT0017167 |
| Rat | rno-miR-299a-5p | MIMAT0000901 |
Stem-loop Details
Rat

Genomic Map