Assay Details
Gene Family ID: | MIPF0000316, mir-467 |
Mature miRNA Sequence: | UGAAUAUACACACACUUACAC |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-669e-3p | MIMAT0017330 |
Stem-loop Details
Mouse

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | mmu-miR-669e* |
---|---|
miRBase Accession Number: | MI0006300 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UGAAUAUACACACACUUACAC |
Species: | Mouse |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000316, mir-467 |
Mature miRNA Sequence: | UGAAUAUACACACACUUACAC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-669e-3p | MIMAT0017330 |