Assay Details
Amplicon Length: | 135 |
Chromosome Location: | Chr. 2 - 10468145 - 10468145 [+] on 10468145 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-669b-5p | MIMAT0003476 |
| Mouse | mmu-miR-669b-3p | MIMAT0017250 |
| Mouse | mmu-miR-669e-3p | MIMAT0017330 |
| Mouse | mmu-miR-669e-5p | MIMAT0005853 |
Stem-loop Details
Mouse

Genomic Map