Assay Details
Gene Family ID: | MIPF0000078, mir-187 | 
Mature miRNA Sequence: | AGGCUACAACACAGGACCCGGG | 
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Mouse | mmu-miR-187-5p | MIMAT0016997 | 
Stem-loop Details
Mouse
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | mmu-miR-187* | 
|---|---|
| miRBase Accession Number: | MI0000229 | 
| miRBase Version: | v22.1 | 
| Mature miRNA Sequence: | AGGCUACAACACAGGACCCGGG | 
| Species: | Mouse | 
| Product Type: | TaqMan™ MicroRNA Assay | 
Gene Family ID: | MIPF0000078, mir-187 | 
Mature miRNA Sequence: | AGGCUACAACACAGGACCCGGG | 
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Mouse | mmu-miR-187-5p | MIMAT0016997 |