Assay Details
Amplicon Length: | 68 |
Chromosome Location: | Chr. 18 - 24429411 - 24429411 [+] on 24429411 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-187-3p | MIMAT0000216 |
| Mouse | mmu-miR-187-5p | MIMAT0016997 |
Stem-loop Details
Mouse

Genomic Map