Assay Details
Gene Family ID: | MIPF0000386, mir-743 |
Mature miRNA Sequence: | UGACUGGCACCAUUCUGGAUAAU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-871-3p | MIMAT0017265 |
Stem-loop Details
Mouse

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | mmu-miR-871-3p |
---|---|
miRBase Accession Number: | MI0005471 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UGACUGGCACCAUUCUGGAUAAU |
Species: | Mouse |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000386, mir-743 |
Mature miRNA Sequence: | UGACUGGCACCAUUCUGGAUAAU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-871-3p | MIMAT0017265 |