Assay Details
Amplicon Length: | 90 |
Chromosome Location: | Chr. X - 66810133 - 66810133 [-] on 66810133 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-871-3p | MIMAT0017265 |
Mouse | mmu-miR-871-5p | MIMAT0004841 |
Stem-loop Details
Mouse
