Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | UGAAUCACACAAAGGCAACUUUU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-377-3p | MIMAT0003123 |
| Cricetulus griseus | cgr-miR-377-3p | MIMAT0023942 |
Stem-loop Details
Rat
Cricetulus griseus

Genomic Map