Assay Details
Amplicon Length: | 60 |
Chromosome Location: | Chr. 6 - 133890944 - 133890944 [-] on 133890944 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-377-5p | MIMAT0017203 |
Rat | rno-miR-377-3p | MIMAT0003123 |
Stem-loop Details
Rat
