Assay Details
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | CAAAAACCGCAAUUACUUUUGCA |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-548h-3p | MIMAT0022723 |
Human | hsa-miR-548z | MIMAT0018446 |
Stem-loop Details
Human
