Assay Details
Amplicon Length: | 98 |
Chromosome Location: | Chr. 8 - 27048653 - 27048653 [-] on 27048653 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548h-5p | MIMAT0005928 |
| Human | hsa-miR-548h-3p | MIMAT0022723 |
Stem-loop Details
Human

Genomic Map