Assay Details
Gene Family ID: | MIPF0000178, mir-208 |
Mature miRNA Sequence: | AAGCUUUUUGCUCGAAUUAUGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-208b-5p | MIMAT0026722 |
| Guinea pig | cpo-miR-208b-5p | MIMAT0047107 |
| Rhesus monkey | mml-miR-208b-5p | MIMAT0026845 |
Stem-loop Details
Human
Guinea pig
Rhesus monkey

Genomic Map